Learn More
Lenz Laborglasinstrumente™ Disposable Ground Joint Sleeve
Fits like a second skin without adding bulk
Brand:  Lenz Laborglasinstrumente™ 01101545
 View more versions of this product
Product Code. 10617153
  • 103.00 € / Pack of 10
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Thin walled
  • Outstanding chemical resistance at operating temperatures from -200 to +260 °C
  • Without grip collar, simplifying use of joint clips
Specifications
Specifications
For ground joints NS 45/40
10
50 μm
Disposable
PTFE
-200 to +260 °C
45/40