missing translation for 'onlineSavingsMsg'
Learn More

Helios Italquartz Dishes for Mahler Bomb Calorimeter, Translucent

Dishes for Mahler Bomb Calorimeter constructed of translucent, durable, resistant, and chemically inert quartz glass.

Brand:  Helios Italquartz 61F00163

 View more versions of this product

Product Code. 15838014

  • 42.60 € / Each
Estimated Shipment: 29-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Translucent
  • Suitable for use in Mahler bomb calorimeter
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications

Specifications

Translucent, 25 mm border
Round
Mahler Bomb Calorimeter
20 mm
Quartz
Evaporating Dish
20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers