missing translation for 'onlineSavingsMsg'
Learn More

Diphosphoryl chloride, 90+%, Thermo Scientific Chemicals

Product Code. 10143344
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13498-14-1
Molecular Formula Cl4O3P2
Molecular Weight (g/mol) 251.76
MDL Number MFCD00061542
InChI Key CNTIXUGILVWVHR-UHFFFAOYSA-N
Synonym diphosphoryl chloride, pyrophosphoryl chloride, diphosphoryl tetrachloride, diphosphoric tetrachloride, pyrophosphoric tetrachloride, dichlorophosphoryl oxyphosphonoyl dichloride, pyrophosphoryl tetrachloride, pyrophosphorylchloride, phosphorodichloridic anhydride
PubChem CID 4432411
SMILES O=P(OP(=O)(Cl)Cl)(Cl)Cl

Specifications

Boiling Point 90.0°C (12.0 mmHg)
Color Colorless through Yellow
Physical Form Liquid
Quantity 25 mL
Assay Percent Range 90+%
Formula Weight 251.76
Percent Purity ≥90%
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.82
Density 1.8200g/mL
Chemical Name or Material Diphosphoryl chloride
Show More Show Less
compliance-icons
Product Identifier
  • Diphosphoryl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.