missing translation for 'onlineSavingsMsg'
Learn More

Dimethylethoxyvinylsilane, 97%, Thermo Scientific Chemicals

Product Code. 11389335
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5356-83-2
Molecular Formula C6H14OSi
Molecular Weight (g/mol) 130.262
MDL Number MFCD00009070
InChI Key JEWCZPTVOYXPGG-UHFFFAOYSA-N
Synonym ethoxydimethylvinylsilane, dimethylethoxyvinylsilane, ethoxydimethyl vinyl silane, silane, ethenylethoxydimethyl, silane, ethoxydimethylvinyl, vinyldimethylethoxysilane, ethenylethoxydimethyl silane, ethoxy dimethyl vinylsilane, dimethylvinylethoxysilane, dimethyl ethenyl silyloxyethane
PubChem CID 79315
IUPAC Name ethenyl-ethoxy-dimethylsilane
SMILES CCO[Si](C)(C)C=C

Specifications

Boiling Point 99°C
Quantity 100 g
UN Number UN1993
Beilstein 1743477
Flash Point −3°C (26°F)
Formula Weight 130.26
Percent Purity 97%
Refractive Index 1.398
Sensitivity Moisture sensitive
Density 0.79
Chemical Name or Material Dimethylethoxyvinylsilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.