missing translation for 'onlineSavingsMsg'
Learn More

Dimethylaminoacetonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11319889
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 926-64-7
Molecular Formula C4H8N2
Molecular Weight (g/mol) 84.12
MDL Number MFCD00001890
InChI Key PLXBWEPPAAQASG-UHFFFAOYSA-N
Synonym dimethylaminoacetonitrile, dimethylamino acetonitrile, acetonitrile, dimethylamino, 2-dimethylamino acetonitrile, n-cyanomethyl dimethylamine, glycinonitrile, n,n-dimethyl, unii-wu34f15n7c, glycinonitrile,n-dimethyl
PubChem CID 61237
IUPAC Name 2-(dimethylamino)acetonitrile
SMILES CN(C)CC#N

Specifications

Density 0.863
Boiling Point 137°C to 138°C
Flash Point 36°C (96°F)
Odor Recognizable
Linear Formula (CH3)2NCH2CN
Refractive Index 1.41
Quantity 100 g
UN Number UN2378
Beilstein 1735677
Formula Weight 84.12
Percent Purity 98%
Chemical Name or Material Dimethylaminoacetonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'