Learn More
Dimethylamine, 2M in methanol, Thermo Scientific Chemicals
42.11 € - 110.55 €
Chemical Identifiers
CAS | 124-40-3 |
---|---|
Molecular Formula | C2H7N |
Molecular Weight (g/mol) | 45.085 |
MDL Number | MFCD00008288 |
InChI Key | ROSDSFDQCJNGOL-UHFFFAOYSA-N |
Synonym | dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution |
PubChem CID | 674 |
ChEBI | CHEBI:17170 |
IUPAC Name | N-methylmethanamine |
SMILES | CNC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11342289
|
Thermo Scientific Alfa Aesar
H27261.AC |
25 mL |
42.11 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11352289
|
Thermo Scientific Alfa Aesar
H27261.AE |
100 mL |
110.55 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dimethylamine is a precursor to several industrially significant compounds. It reacts with carbon disulfide to give dimethyl dithiocarbamate, a precursor to a family of chemicals widely used in the vulcanization of rubber. It is raw material for the production of many agrichemicals and pharmaceuticals, such as dimefox and diphenhydramine, respectively. The surfactant lauryl dimethylamine oxide is found in soaps and cleaning compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethylamine is a precursor to several industrially significant compounds. It reacts with carbon disulfide to give dimethyl dithiocarbamate, a precursor to a family of chemicals widely used in the vulcanization of rubber. It is raw material for the production of many agrichemicals and pharmaceuticals, such as dimefox and diphenhydramine, respectively. The surfactant lauryl dimethylamine oxide is found in soaps and cleaning compounds.
Solubility
Fully miscible in water.
Notes
Hygroscopic. Temperature sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, water, moisture, mercury, chlorine, copper and copper compounds.
Chemical Identifiers
124-40-3 | |
45.085 | |
ROSDSFDQCJNGOL-UHFFFAOYSA-N | |
674 | |
N-methylmethanamine |
C2H7N | |
MFCD00008288 | |
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution | |
CHEBI:17170 | |
CNC |
Specifications
124-40-3 | |
5°C (41°F) | |
(CH3)2NH | |
MFCD00008288 | |
605257 | |
14,3228 | |
Fully miscible in water. | |
CNC | |
45.085 | |
CHEBI:17170 | |
2M in methanol |
0.775 | |
C2H7N | |
1.335 | |
UN3286 | |
Hygroscopic | |
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution | |
ROSDSFDQCJNGOL-UHFFFAOYSA-N | |
N-methylmethanamine | |
674 | |
45.08 | |
Dimethylamine |
Safety and Handling
GHS H Statement
H225-H311-H331-H370-H314
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P307+P311-P310-P311-P312-P330-P363-P370+P378q-P501c
H225-H301+H311+H331-H318-H335-H370
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 204-697-4
missing translation for 'rtecsNumber' : IP8750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.