missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide, Multi-Compendial, J.T. Baker™
Click to view available options
Quantity:
100 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Bottle
Specifications
Specifications
Quantity | 4 L |
Flash Point | 89.00°C |
Packaging | Narrow Mouth PVC-coated Amber Bottle |
Specific Gravity | 1L = 1.10kg |
IUPAC Name | methylsulfinylmethane |
Formula Weight | 78.13 |
Grade | Multi-Compendial |
Chemical Name or Material | Dimethyl Sulfoxide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction