missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl Sulfoxide, Fisher BioReagents™ Product Code.: 11463883

Dimethyl Sulfoxide, Fisher BioReagents™

Product Code. 11463883
4 L, HDPE Bottle
Click to view available options
Quantity:
100 mL
1 L
4 L
Packaging:
Glass Bottle
HDPE Bottle
This item is not returnable. View return policy

Product Code. 11463883

Brand: Fisher BioReagents BP2314

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Dimethyl Sulfoxide, >-99.7%, C2H6OS, CAS Number-67-68-5, methane, sulfinylbis, dmso, demasorb, dimethyl sulphoxide, demavet, dimethyl sulfoxide, demsodrox, dimethylsulfoxide, dimexide, methyl sulfoxide, 100mL, <-0.30 Absorbance Units at 280nm, Vapor Pressure: 0.55mbar at 20 deg.C, >-99.7 %

Dimethyl Sulfoxide, >-99.7%, C2H6OS, CAS Number-67-68-5, methane, sulfinylbis, dmso, demasorb, dimethyl sulphoxide, demavet, dimethyl sulfoxide, demsodrox, dimethylsulfoxide, dimexide, methyl sulfoxide, 100mL, <-0.30 Absorbance Units at 280nm, Vapor Pressure: 0.55mbar at 20 deg.C, >-99.7 %

DMSO is a cryopreservation agent for cells, and may also be used in bacterial transformations.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Absorbance ≤0.30 Absorbance Units at 280nm
Color Colorless
Decomposition Information >190°C
Density 1.100g/cm³
Boiling Point 189°C
Quantity 4 L
Identification Pass Test
Packaging HDPE Bottle
Physical Form Liquid
Assay Percent Range ≥99.7 %
Residue after Evaporation 0.001% max.
Linear Formula (CH3)2SO
Vapor Pressure 0.55mbar at 20°C
Viscosity 1.98 mPa/s at 25°C
Water 0.2% max.
Merck Index 15, 3285
Formula Weight 78.13
Percent Purity ≥99.7%
Chemical Name or Material Dimethyl Sulfoxide
Show More Show Less
Product Identifier
  • Methyl sulfoxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl Sulfoxide, Fisher BioReagents™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.