missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl sulfOxide, ≥99.9% (GC), Honeywell Riedel-de Haën™
CHROMASOLV™, GC-Headspace tested,≥99.9%
33.74 € - 513.29 €
Chemical Identifiers
CAS | 67-68-5 |
---|---|
Molecular Formula | C2H6OS |
Molecular Weight (g/mol) | 78.13 |
MDL Number | MFCD00002089 |
InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide |
PubChem CID | 679 |
ChEBI | CHEBI:28262 |
SMILES | CS(C)=O |
Description
Honeywell Chromasolv™ GC-headspace solvents:
- developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
- very pure solvents with extremely low concentrations of the defined residual solvents
- state-of-the art production and application tests
Chemical Identifiers
67-68-5 | |
78.13 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
679 | |
CS(C)=O |
C2H6OS | |
MFCD00002089 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CHEBI:28262 |
Specifications
67-68-5 | |
42%, 63°F | |
573°F | |
(CH3)2SO | |
0.42 mmHg (20°C) | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CS(C)=O | |
78.13 | |
CHEBI:28262 | |
≥99.9% | |
Dimethyl sulfoxide |
1.10g/mL | |
87°C (188.6°F) | |
C2H6OS | |
MFCD00002089 | |
NA1993 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
methanesulfinylmethane | |
679 | |
78.13 | |
CHROMASOLV™ |
Safety and Handling
missing translation for 'dotInformation' : NA 1993 / PGIII
missing translation for 'rtecsNumber' : 200-664-3