missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxymethylsilane, 97%, Thermo Scientific Chemicals

Product Code. 11302937
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16881-77-9
Molecular Formula C3H9O2Si
Molecular Weight (g/mol) 105.188
MDL Number MFCD00048019
InChI Key PKTOVQRKCNPVKY-UHFFFAOYSA-N
Synonym methyldimethoxysilane, dimethoxy methyl silane, silane, dimethoxymethyl, dimethoxy methyl silicon, methyl dimethoxysilane, c3h10o2si, methyl dimethoxy silane, dimethoxy methyl silane #, dimethoxy methyl silyl
PubChem CID 6329250
IUPAC Name dimethoxy(methyl)silicon
SMILES CO[Si](C)OC

Specifications

Boiling Point 61°C
Melting Point -136°C
Quantity 25 g
UN Number UN1993
Beilstein 2035898
Flash Point 2°C (35°F)
Formula Weight 106.2
Percent Purity 97%
Refractive Index 1.36
Sensitivity Moisture sensitive
Density 0.861
Chemical Name or Material Dimethoxymethylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'