Learn More
Dilithium tetrabromonickelate(II), 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
81.60 € - 255.00 €
Chemical Identifiers
CAS | 13826-95-4, 109-99-9 |
---|---|
Molecular Formula | Br4Li2Ni |
Molecular Weight (g/mol) | 392.19 |
InChI Key | AVJDNPHXGKKFIA-UHFFFAOYSA-J |
Synonym | di-lithium tetrabromonickelate ii, agn-pc-0nf0dx, lithium tetrabromonickelate ii solution, dilithium tetrabromonickelate ii solution |
PubChem CID | 11047451 |
IUPAC Name | dilithium;tetrabromonickel(2-) |
SMILES | [Li+].[Li+].[Ni-2](Br)(Br)(Br)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11954341
|
Thermo Scientific Acros
434541000 |
100 mL | AcroSeal™ Glass Bottle |
81.60 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11964341
|
Thermo Scientific Acros
434548000 |
800 mL | AcroSeal™ Glass Bottle |
255.00 €
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13826-95-4, 109-99-9 | |
392.19 | |
di-lithium tetrabromonickelate ii, agn-pc-0nf0dx, lithium tetrabromonickelate ii solution, dilithium tetrabromonickelate ii solution | |
dilithium;tetrabromonickel(2-) |
Br4Li2Ni | |
AVJDNPHXGKKFIA-UHFFFAOYSA-J | |
11047451 | |
[Li+].[Li+].[Ni-2](Br)(Br)(Br)Br |
Specifications
13826-95-4, 109-99-9 | |
Solution | |
Li2NiBr4 | |
di-lithium tetrabromonickelate ii, agn-pc-0nf0dx, lithium tetrabromonickelate ii solution, dilithium tetrabromonickelate ii solution | |
[Li+].[Li+].[Ni-2](Br)(Br)(Br)Br | |
392.19 | |
392.19 | |
1.0650g/mL |
Blue-Green to Brown | |
Br4Li2Ni | |
−18°C | |
AVJDNPHXGKKFIA-UHFFFAOYSA-J | |
dilithium;tetrabromonickel(2-) | |
11047451 | |
1.065 | |
Dilithium tetrabromonickelate(II) |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
May cause cancer.
May form explosive peroxides.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
GHS Signal Word: Danger
- Dilithium tetrabromonickelate(II), 0.5M solution in THF
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H350-May cause cancer.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only