Learn More
Diglycolic acid, 98%, Thermo Scientific Chemicals
27.50 € - 235.00 €
Chemical Identifiers
CAS | 110-99-6 |
---|---|
Molecular Formula | C4H6O5 |
Molecular Weight (g/mol) | 134.09 |
MDL Number | MFCD00004309 |
InChI Key | QEVGZEDELICMKH-UHFFFAOYSA-N |
Synonym | diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid |
PubChem CID | 8088 |
IUPAC Name | 2-(carboxymethoxy)acetic acid |
SMILES | OC(=O)COCC(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11480198
|
Thermo Scientific Alfa Aesar
B23758.14 |
25 g |
27.50 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490198
|
Thermo Scientific Alfa Aesar
B23758.22 |
100 g |
63.70 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400208
|
Thermo Scientific Alfa Aesar
B23758.36 |
500 g |
235.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Diglycolic acid was used in the preparation of needle-like aragonite particles from calcium nitrate aqueous solutions. It is also used in the synthesis of N-interlinked imipramines which show apoptotic activity against malignant cells as well as signs of combating Burkitt’s lymphoma.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiglycolic acid was used in the preparation of needle-like aragonite particles from calcium nitrate aqueous solutions. It is also used in the synthesis of N-interlinked imipramines which show apoptotic activity against malignant cells as well as signs of combating Burkitt’s lymphoma.
Solubility
Soluble in water, methanol.
Notes
Store in cool, dry conditions in well sealed containers. Store away from oxidizing agents.
Chemical Identifiers
110-99-6 | |
134.09 | |
QEVGZEDELICMKH-UHFFFAOYSA-N | |
8088 | |
OC(=O)COCC(O)=O |
C4H6O5 | |
MFCD00004309 | |
diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid | |
2-(carboxymethoxy)acetic acid |
Specifications
110-99-6 | |
C4H6O5 | |
1072117 | |
Soluble in water,methanol. | |
OC(=O)COCC(O)=O | |
134.09 | |
134.09 | |
Diglycolic acid |
140°C to 145°C | |
MFCD00004309 | |
diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid | |
QEVGZEDELICMKH-UHFFFAOYSA-N | |
2-(carboxymethoxy)acetic acid | |
8088 | |
98% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 203-823-5
missing translation for 'rtecsNumber' : AJ1530000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only