Learn More
Karl Hecht™ Assistent™ Digital Interval Timers
Excellent for various timing purposes in the laboratory. Karl Hecht™ Assistent™ Digital Interval Timers are programmable up to 99 minutes and 59 seconds.
Brand:  Karl Hecht™ 41977060
 View more versions of this product
Product Code. 10247381
  • 33.00 € / Each
Explore more special offers
Description
Description
  • With magnetic back
  • Includes battery
  • Counts down in seconds
  • Memory recall function
  • Individual setting for minutes/seconds
  • LCD display, 4 digits showing minutes/seconds
  • Alarm sounds for 30 seconds when countdown reaches zero
Specifications
Specifications
Digital Timer
84 x 77 x 14 mm
14 mm
Digital
14 mm