Learn More
Diethylzinc, nominally 15% w/w in hexanes, Thermo Scientific™
62.40 €
Chemical Identifiers
CAS | 557-20-0 |
---|---|
Molecular Formula | C4H10Zn |
Molecular Weight (g/mol) | 123.50 |
MDL Number | MFCD00009021 |
InChI Key | HQWPLXHWEZZGKY-UHFFFAOYSA-N |
Synonym | diethylzinc, zinc, diethyl, diethyl zinc, zinc ethide, zinc ethyl, unii-s0w5nqh7c6, et2zn, znet2, s0w5nqh7c6, c2h5 2zn |
PubChem CID | 11185 |
SMILES | CC[Zn]CC |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
557-20-0 | |
123.50 | |
HQWPLXHWEZZGKY-UHFFFAOYSA-N | |
11185 |
C4H10Zn | |
MFCD00009021 | |
diethylzinc, zinc, diethyl, diethyl zinc, zinc ethide, zinc ethyl, unii-s0w5nqh7c6, et2zn, znet2, s0w5nqh7c6, c2h5 2zn | |
CC[Zn]CC |
Specifications
557-20-0 | |
0.720g/mL | |
C4H10Zn | |
MFCD00009021 | |
Air & Moisture Sensitive | |
HQWPLXHWEZZGKY-UHFFFAOYSA-N | |
diethylzinc | |
11185 | |
Liquid |
Brown to Colorless | |
−40°C (−40°F) | |
Zn(C2H5)2 | |
UN3394 | |
diethylzinc, zinc, diethyl, diethyl zinc, zinc ethide, zinc ethyl, unii-s0w5nqh7c6, et2zn, znet2, s0w5nqh7c6, c2h5 2zn | |
CC[Zn]CC | |
123.50 | |
123.49 | |
Diethylzinc |
Safety and Handling
GHS H Statement
H225-H250-H260-H252-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
missing translation for 'tsca' : TSCA
Your input is important to us. Please complete this form to provide feedback related to the content on this product.