Learn More
Diethylsilane, 96+%, Thermo Scientific Chemicals
63.01 € - 226.13 €
Chemical Identifiers
CAS | 542-91-6 |
---|---|
Molecular Formula | C4H10Si |
Molecular Weight (g/mol) | 86.209 |
MDL Number | MFCD00039877 |
InChI Key | KZZFGAYUBYCTNX-UHFFFAOYSA-N |
Synonym | diethylsilane, silane, diethyl, unii-t6ea843e4k, c2h5 2sih2, 3-silapentane, silane, diethyl-,, diethylsilylene radical, diethylsilane, high purity min |
PubChem CID | 6327565 |
IUPAC Name | diethylsilicon |
SMILES | CC[Si]CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11359175
|
Thermo Scientific Alfa Aesar
L16468.06 |
5 g |
63.01 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11369175
|
Thermo Scientific Alfa Aesar
L16468.14 |
25 g |
226.13 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Diethylsilane is used as a precursor to prepare (3-allylsulfanyl-propyl)-diethyl-silane by using (Ph3P)3RhCl as reagent. It is also used in the chemical vapor deposition of SiO2 for microelectronics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethylsilane is used as a precursor to prepare (3-allylsulfanyl-propyl)-diethyl-silane by using (Ph3P)3RhCl as reagent. It is also used in the chemical vapor deposition of SiO2 for microelectronics.
Solubility
Miscible with terahydrofuran, diethyl ether, 1,4-dioxane and benzene.
Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, bases, oxygen and water.
Chemical Identifiers
542-91-6 | |
86.209 | |
KZZFGAYUBYCTNX-UHFFFAOYSA-N | |
6327565 | |
CC[Si]CC |
C4H10Si | |
MFCD00039877 | |
diethylsilane, silane, diethyl, unii-t6ea843e4k, c2h5 2sih2, 3-silapentane, silane, diethyl-,, diethylsilylene radical, diethylsilane, high purity min | |
diethylsilicon |
Specifications
56°C | |
542-91-6 | |
MFCD00039877 | |
1730902 | |
14,3128 | |
Miscible with terahydrofuran,diethyl ether,1,4-dioxane and benzene. | |
CC[Si]CC | |
86.209 | |
88.23 | |
Mild | |
Moisture sensitive | |
Diethylsilane |
-132°C | |
C4H10Si | |
UN1993 | |
−20°C (−4°F) | |
diethylsilane, silane, diethyl, unii-t6ea843e4k, c2h5 2sih2, 3-silapentane, silane, diethyl-,, diethylsilylene radical, diethylsilane, high purity min | |
KZZFGAYUBYCTNX-UHFFFAOYSA-N | |
diethylsilicon | |
6327565 | |
≥98% | |
1.392 | |
0.684 |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 208-834-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only