missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Monoethyl Ether, ReagentPlus™ 99%, Honeywell™

Product Code. 15675780
Click to view available options
Quantity:
100 mL
1 L
4 L
20 L
Packaging:
Glass bottle
Stainless steel drum
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-90-0
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.175
MDL Number MFCD00002872
InChI Key XXJWXESWEXIICW-UHFFFAOYSA-N
Synonym diethylene glycol monoethyl ether, 2-2-ethoxyethoxy ethanol, carbitol, transcutol, ethoxy diglycol, dioxitol, ethyl carbitol, ethyl digol, carbitol solvent, ethanol, 2-2-ethoxyethoxy
PubChem CID 8146
ChEBI CHEBI:40572
IUPAC Name 2-(2-ethoxyethoxy)ethanol
SMILES CCOCCOCCO

Specifications

Density 0.999g/mL at 25°C
Packaging Glass bottle
Linear Formula C2H5OCH2CH2OCH2CH2OH
Refractive Index 1.427
Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 1736441
Vapor Density 4.63 (vs air)
Formula Weight 134.17g/mol
Percent Purity 99%
Chemical Name or Material Diethylene glycol monoethyl ether
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'