missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine, Certified AR for Analysis, Fisher Chemical™

94.10 € - 158.00 €

Chemical Identifiers

CAS 109-89-7
MDL Number 9032
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10345990
View Documents
Fisher Chemical
D/2110/08
500 mL
94.10 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10408790
View Documents
Fisher Chemical
D/2110/17
2.5 L
158.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

109-89-7
9032

Specifications

109-89-7
Colorless
55°C
C4H11N
73.13g/mol
-50°C
12
Amber glass bottle
9032
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Diethylamine

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.