missing translation for 'onlineSavingsMsg'
Learn More

Diethyl n-butylmalonate, 99%, Thermo Scientific™

Product Code. 11301947
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133-08-4
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.28
MDL Number MFCD00009170
InChI Key RPNFNBGRHCUORR-UHFFFAOYSA-N
Synonym diethyl butylmalonate, diethyl n-butylmalonate, butylmalonic acid diethyl ester, diethyl 2-butylmalonate, ethyl butylmalonate, propanedioic acid, butyl-, diethyl ester, unii-cyy2q744wb, cyy2q744wb, n-butylmalonic acid diethyl ester, malonic acid, butyl-, diethyl ester
PubChem CID 67244
IUPAC Name diethyl 2-butylpropanedioate
SMILES CCCCC(C(=O)OCC)C(=O)OCC

Specifications

Density 0.982
Boiling Point 236°C to 239°C
Flash Point 93°C (199°F)
Refractive Index 1.423
Quantity 500 mL
Beilstein 608785
Merck Index 14,1576
Formula Weight 216.28
Percent Purity 99%
Chemical Name or Material Diethyl n-butylmalonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.