missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 1,3-acetonedicarboxylate, 95%, Thermo Scientific Chemicals
60.00 € - 199.00 €
Chemical Identifiers
CAS | 105-50-0 |
---|---|
Molecular Formula | C9H14O5 |
Molecular Weight (g/mol) | 202.21 |
MDL Number | MFCD00009200 |
InChI Key | ZSANYRMTSBBUCA-UHFFFAOYSA-N |
Synonym | diethyl 1,3-acetonedicarboxylate, diethyl 3-oxoglutarate, 1,5-diethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, diethyl ester, ethyl acetonedicarboxylate, ethyl 3-oxoglutarate, diethyl acetone-1,3-dicarboxylate, diethyl acetone dicarboxylate, 3-oxopentanedioic acid diethyl ester, acetonedicarboxylic acid diethyl ester |
PubChem CID | 66045 |
IUPAC Name | diethyl 3-oxopentanedioate |
SMILES | CCOC(=O)CC(=O)CC(=O)OCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10309420
|
Thermo Scientific Acros
325630500 |
50 mL | Glass bottle |
60.00 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10636301
|
Thermo Scientific Acros
325632500 |
250 mL | Glass bottle |
199.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
105-50-0 | |
202.21 | |
ZSANYRMTSBBUCA-UHFFFAOYSA-N | |
66045 | |
CCOC(=O)CC(=O)CC(=O)OCC |
C9H14O5 | |
MFCD00009200 | |
diethyl 1,3-acetonedicarboxylate, diethyl 3-oxoglutarate, 1,5-diethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, diethyl ester, ethyl acetonedicarboxylate, ethyl 3-oxoglutarate, diethyl acetone-1,3-dicarboxylate, diethyl acetone dicarboxylate, 3-oxopentanedioic acid diethyl ester, acetonedicarboxylic acid diethyl ester | |
diethyl 3-oxopentanedioate |
Specifications
105-50-0 | |
1.1120g/mL | |
86°C | |
94% min. (GC) | |
C2H5O2CCH2COCH2CO2C2H5 | |
MFCD00009200 | |
1.112 | |
Solubility in water: 10g/L in water (20°C) | |
CCOC(=O)CC(=O)CC(=O)OCC | |
202.21 | |
202.21 | |
Liquid |
Colorless to Yellow | |
250.0°C | |
Authentic | |
C9H14O5 | |
1.4380 to 1.4420 | |
03, 791 | |
diethyl 1,3-acetonedicarboxylate, diethyl 3-oxoglutarate, 1,5-diethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, diethyl ester, ethyl acetonedicarboxylate, ethyl 3-oxoglutarate, diethyl acetone-1,3-dicarboxylate, diethyl acetone dicarboxylate, 3-oxopentanedioic acid diethyl ester, acetonedicarboxylic acid diethyl ester | |
ZSANYRMTSBBUCA-UHFFFAOYSA-N | |
diethyl 3-oxopentanedioate | |
66045 | |
95% | |
Diethyl 1, 3-acetonedicarboxylate, 95% |
Safety and Handling
MOISTURE SENSITIVE
missing translation for 'einecsNumber' : 203-302-2
missing translation for 'tsca' : TSCA
RUO – Research Use Only