missing translation for 'onlineSavingsMsg'
Learn More

Diethoxydiphenylsilane, 98%, Thermo Scientific Chemicals

29.00 € - 346.73 €

Chemical Identifiers

CAS 2553-19-7
Molecular Formula C16H20O2Si
Molecular Weight (g/mol) 272.42
MDL Number MFCD00015126
InChI Key ZZNQQQWFKKTOSD-UHFFFAOYSA-N
Synonym diphenyldiethoxysilane, silane, diethoxydiphenyl, diethoxy diphenyl silane, benzene, 1,1'-diethoxysilylene bis, acmc-1cmgu, diethoxy-di phenyl silane, diethoxydiphenylsilane, zznqqqwfkktosd-uhfffaoysa
PubChem CID 75705
IUPAC Name diethoxy(diphenyl)silane
SMILES CCO[Si](OCC)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11465826
View Documents
Thermo Scientific Alfa Aesar
B22204.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11475826
View Documents
Thermo Scientific Alfa Aesar
B22204.18
50 g
100.30 €
50g
Please to purchase this item. Need a web account? Register with us today!
11485826
View Documents
Thermo Scientific Alfa Aesar
B22204.30
250 g
346.73 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reagent for preparation of diphenylsilylene derivatives of diols by exchange.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2553-19-7
272.42
ZZNQQQWFKKTOSD-UHFFFAOYSA-N
75705
CCO[Si](OCC)(C1=CC=CC=C1)C1=CC=CC=C1
C16H20O2Si
MFCD00015126
diphenyldiethoxysilane, silane, diethoxydiphenyl, diethoxy diphenyl silane, benzene, 1,1'-diethoxysilylene bis, acmc-1cmgu, diethoxy-di phenyl silane, diethoxydiphenylsilane, zznqqqwfkktosd-uhfffaoysa
diethoxy(diphenyl)silane

Specifications

166°C to 168°C (15mmHg)
C16H20O2Si
2281302
diphenyldiethoxysilane, silane, diethoxydiphenyl, diethoxy diphenyl silane, benzene, 1,1'-diethoxysilylene bis, acmc-1cmgu, diethoxy-di phenyl silane, diethoxydiphenylsilane, zznqqqwfkktosd-uhfffaoysa
CCO[Si](OCC)(C1=CC=CC=C1)C1=CC=CC=C1
272.42
272.42
1.5255
1.033
2553-19-7
MFCD00015126
175°C (347°F)
ZZNQQQWFKKTOSD-UHFFFAOYSA-N
diethoxy(diphenyl)silane
75705
98%
Moisture sensitive
Diethoxydiphenylsilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 219-860-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.