Learn More
Dicyclopentanylmethyl acrylate, Thermo Scientific Chemicals
27.60 € - 306.00 €
Chemical Identifiers
CAS | 93962-84-6 |
---|---|
Molecular Formula | C14H20O2 |
Molecular Weight (g/mol) | 220.312 |
MDL Number | MFCD22380704 |
InChI Key | PMPNHSZLJPXGCD-UHFFFAOYSA-N |
Synonym | octahydro-4,7-methano-1h-indenyl methyl acrylate, acmc-20mslm, tricyclo 5.2.1.0 2 ,? decan-3-ylmethyl prop-2-enoate |
PubChem CID | 22051805 |
SMILES | C=CC(=O)OCC1CCC2C1C3CCC2C3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15462737
|
Thermo Scientific Alfa Aesar
046652.06 |
5 g |
27.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15472737
|
Thermo Scientific Alfa Aesar
046652.14 |
25 g |
86.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15482737
|
Thermo Scientific Alfa Aesar
046652.22 |
100 g |
306.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
93962-84-6 | |
220.312 | |
PMPNHSZLJPXGCD-UHFFFAOYSA-N | |
22051805 |
C14H20O2 | |
MFCD22380704 | |
octahydro-4,7-methano-1h-indenyl methyl acrylate, acmc-20mslm, tricyclo 5.2.1.0 2 ,? decan-3-ylmethyl prop-2-enoate | |
C=CC(=O)OCC1CCC2C1C3CCC2C3 |
Specifications
93962-84-6 | |
C14H20O2 | |
octahydro-4,7-methano-1h-indenyl methyl acrylate, acmc-20mslm, tricyclo 5.2.1.0 2 ,? decan-3-ylmethyl prop-2-enoate | |
C=CC(=O)OCC1CCC2C1C3CCC2C3 | |
22051805 | |
Liquid |
Yellow | |
MFCD22380704 | |
PMPNHSZLJPXGCD-UHFFFAOYSA-N | |
220.312 | |
220.31 | |
Dicyclopentanylmethyl acrylate |
Safety and Handling
GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
H315-H317-H319-H335
missing translation for 'einecsNumber' : 300-723-4
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only