missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexyltin dibromide, Thermo Scientific™

96.00 € - 318.00 €

Chemical Identifiers

CAS 2954-94-1
Molecular Formula C12H22Br2Sn
Molecular Weight (g/mol) 444.826
MDL Number MFCD00015370
InChI Key RHFTTZVNBAHQBF-UHFFFAOYSA-L
Synonym dicyclohexyltin dibromide, stannane, dibromodicyclohexyl, dibromo dicyclohexyl stannane, dicyclohexyltindibromide, stannane,dibromodicyclohexyl
PubChem CID 97477
IUPAC Name dibromo(dicyclohexyl)stannane
SMILES C1CCC(CC1)[Sn](C2CCCCC2)(Br)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11365039
View Documents
Thermo Scientific Alfa Aesar
087800.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11375039
View Documents
Thermo Scientific Alfa Aesar
087800.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2954-94-1
444.826
RHFTTZVNBAHQBF-UHFFFAOYSA-L
97477
C1CCC(CC1)[Sn](C2CCCCC2)(Br)Br
C12H22Br2Sn
MFCD00015370
dicyclohexyltin dibromide, stannane, dibromodicyclohexyl, dibromo dicyclohexyl stannane, dicyclohexyltindibromide, stannane,dibromodicyclohexyl
dibromo(dicyclohexyl)stannane

Specifications

2954-94-1
White
C12H22Br2Sn
UN3146
dicyclohexyltin dibromide, stannane, dibromodicyclohexyl, dibromo dicyclohexyl stannane, dicyclohexyltindibromide, stannane,dibromodicyclohexyl
C1CCC(CC1)[Sn](C2CCCCC2)(Br)Br
444.826
444.82
Dicyclohexyltin dibromide
58°C to 64°C
∼150°C (0.3mmHg)
MFCD00015370
Moisture sensitive
RHFTTZVNBAHQBF-UHFFFAOYSA-L
dibromo(dicyclohexyl)stannane
97477
Crystalline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.