missing translation for 'onlineSavingsMsg'
Learn More
Learn More
μ-Dichlorotetraethylene dirhodium(I), Thermo Scientific Chemicals
96.70 € - 699.00 €
Chemical Identifiers
CAS | 12081-16-2 |
---|---|
Molecular Formula | C8H16Cl2Rh2 |
Molecular Weight (g/mol) | 388.93 |
MDL Number | MFCD00013206 |
InChI Key | OOQJCPOXJFCGCR-UHFFFAOYSA-L |
Synonym | chlorobis ethylene rhodium i dimer, dichlorotetra ethylene dirhodium i |
PubChem CID | 45357812 |
IUPAC Name | ethene;rhodium;dichloride |
SMILES | Cl[Rh].Cl[Rh].C=C.C=C.C=C.C=C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10144120
|
Thermo Scientific Acros
197591000 |
100 mg | Glass bottle |
96.70 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10026093
|
Thermo Scientific
197590010 |
1 g | Glass bottle |
699.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12081-16-2 | |
388.93 | |
OOQJCPOXJFCGCR-UHFFFAOYSA-L | |
45357812 | |
Cl[Rh].Cl[Rh].C=C.C=C.C=C.C=C |
C8H16Cl2Rh2 | |
MFCD00013206 | |
chlorobis ethylene rhodium i dimer, dichlorotetra ethylene dirhodium i | |
ethene;rhodium;dichloride |
Specifications
12081-16-2 | |
Authentic | |
MFCD00013206 | |
Solubility in water: insoluble | |
Cl[Rh].Cl[Rh].C=C.C=C.C=C.C=C | |
388.93 | |
388.92 | |
µ-Dichlorotetraethylene dirhodium(I) |
Rust | |
C8H16Cl2Rh2 | |
chlorobis ethylene rhodium i dimer, dichlorotetra ethylene dirhodium i | |
OOQJCPOXJFCGCR-UHFFFAOYSA-L | |
ethene;rhodium;dichloride | |
45357812 | |
Crystalline Powder and/or Chunks |
Safety and Handling
AIR SENSITIVE
missing translation for 'einecsNumber' : 235-145-0
RUO – Research Use Only