missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethyl-n-octylsilane, 98%, Thermo Scientific Chemicals

Product Code. 11379235
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14799-93-0
Molecular Formula C9H20Cl2Si
Molecular Weight (g/mol) 227.24
MDL Number MFCD00042606
InChI Key QHBMMABVNRSRHW-UHFFFAOYSA-N
Synonym n-octylmethyldichlorosilane, silane, dichloromethyloctyl, dichloro methyl-n-octylsilane, methyloctyldichlorosilane, dichloro methyl octyl silane, dichloro methyl octylsilane, octylmethyldichlorosilane, acmc-209czn, methyloctyl dichlorosilane, dichloro methyl octylsilane #
PubChem CID 84649
IUPAC Name dichloro-methyl-octylsilane
SMILES CCCCCCCC[Si](C)(Cl)Cl

Specifications

Boiling Point 224°C to 226°C
Quantity 10 g
UN Number UN2987
Beilstein 1740697
Flash Point 98°C (208°F)
Formula Weight 227.25
Percent Purity 98%
Chemical Name or Material Dichloromethyl-n-octylsilane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'