missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, Spectrophotometric Grade, 99.7+%, stab. with amylene, Thermo Scientific Chemicals

Product Code. 11388367
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Dichloromethane is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -95°C
Density 1.325 g/mL
Boiling Point 39°C to 40°C
Quantity 4 L
Odor Sweet
Physical Form Liquid
Refractive Index 1.4244
UN Number UN1593
Beilstein 1730800
Merck Index 14,6063
Solubility Information Miscible with ethyl acetate,alcohol,hexanes,methanol,diethyl ether,n-octanol,acetone benzene,carbon tetrachloride,diethyl ether and chloroform. Immiscible with water.
Formula Weight 84.93
Percent Purity ≥99.7%
Grade Spectrophotometric
Chemical Name or Material Dichloromethane, Stabilized with amylene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'