missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, Gel Permeation Chromatography (GPC), Stabilised with Amylene, Fisher Chemical

Product Code. 10715331
Click to view available options
2.5L, Plastic Coated Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point -97°C
CAS 75-09-2
Color Colorless
Boiling Point 39°C
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Molecular Formula CH2Cl2
MDL Number MFCD00000881
Vapor Pressure 350mbar at 20°C
Viscosity 0.43 mPaS at 20°C
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES ClCCl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
Formula Weight 84.93g/mol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'