missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, Extra Dry, Stabilised with Amylene, for Organic Synthesis, Fisher Chemical

Product Code. 10375890
Click to view available options
1L, Plastic Coated Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point -97°C
CAS 75-09-2
Color Colorless
Boiling Point 39°C
Quantity 1 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Molecular Formula CH2Cl2
MDL Number MFCD00000881
Vapor Pressure 350mbar at 20°C
Viscosity 0.43 mPaS at 20°C
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES ClCCl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
Formula Weight 84.93g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'