missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane-d{2}, 99.9% (Isotopic), Thermo Scientific Chemicals

Product Code. 11374418
Click to view available options
Quantity:
10 Ea.
2 Ea.
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Acts as a solvent in Nuclear magnetic resonance (NMR) spectroscopy.

Dichloromethane-d{2} acts as a solvent in Nuclear magnetic resonance (NMR) spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1665-00-5
MDL Number MFCD00000882
Synonym dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d

Specifications

Density 1.35
Packaging Prescored ampoules, 0.75mL/ampoule
Quantity 10 Ea.
Sensitivity Moisture sensitive
Solubility Information Miscible with alcohol,ether and dimethyl formamide.
Formula Weight 86.95
Percent Purity 99.9%
Physical Form Liquid
Chemical Name or Material Dichloromethane-d2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.