missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane CHROMASOLV™, for HPLC, ≥99.8%, contains amylene as stabilizer, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
100 mL
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
Melting Point | -97°C |
Density | 1.325g/mL at 25°C |
Boiling Point | 39.8°C to 40°C |
Quantity | 100 mL |
Refractive Index | n20/D 1.424 |
Linear Formula | CH2Cl2 |
Vapor Pressure | 6.83 psi (20°C) |
Beilstein | 1730800 |
Formula Weight | 84.93g/mol |
Percent Purity | ≥99.8% |
Show More |
Safety and Handling
Product Identifier
- Dichloromethane
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'