missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, ACS Reagent, contains 40-150 amylene as stabilizer, ≥99.5%, Honeywell Riedel-de Haën™
ACS Reagent, contains 40-150 amylene as stabilizer,≥99.5%
57.27 € - 757.24 €
Chemical Identifiers
CAS | 75-09-2 |
---|---|
Molecular Formula | CH2Cl2 |
Molecular Weight (g/mol) | 84.93 |
MDL Number | MFCD00000881 |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
PubChem CID | 6344 |
ChEBI | CHEBI:15767 |
IUPAC Name | dichloromethane |
SMILES | ClCCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15677830
|
Honeywell-Riedel-de Haen
D65100-1L |
1 L | Glass Bottle | N/A | |||||
15687830
|
Honeywell-Riedel-de Haen
D65100-2.5L |
2.5 L | Glass Bottle | N/A | |||||
15607840
|
Honeywell-Riedel-de Haen
D65100-6X1L |
6 x 1 L | Glass Bottle | N/A | |||||
15667830
|
Honeywell-Riedel-de Haen
D65100-10L |
10 L | Stainless Steel Can | N/A | |||||
15697830
|
Honeywell-Riedel-de Haen
D65100-20L |
20 L | Stainless Steel Can | N/A | |||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
75-09-2 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
6344 | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
CHEBI:15767 | |
ClCCl |
Specifications
-97°C (literature) | |
≥99.0000% | |
1.325 g/mL at 25 °C(lit.) | |
CH2Cl2 | |
MFCD00000881 | |
1730800 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
ClCCl | |
84.93 | |
CHEBI:15767 | |
>99.5% | |
Dichloromethane, ACS Reagent, contains 40-150 amylene as stabilizer, ≥99.5% |
75-09-2 | |
≥99.0000% | |
39°C to 41°C | |
CH2Cl2 | |
UN1593 | |
max. 0.02% | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
6344 | |
84.93 | |
ACS Reagent |
Safety and Handling
missing translation for 'shelfLife' : 1260 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Dichloromethane
missing translation for 'einecsNumber' : 200-838-9
missing translation for 'rtecsNumber' : PA8050000
Recommended Storage : Room Temp