Learn More
Dichloroisocyanuric acid sodium salt monohydrate, 97%, Thermo Scientific Chemicals
34.77 € - 163.82 €
Chemical Identifiers
CAS | 52671-45-1 |
---|---|
Molecular Formula | C3Cl2N3NaO3·H2O |
Molecular Weight (g/mol) | 237.96 |
InChI Key | FIHQYHSNTSJNGG-UHFFFAOYSA-N |
Synonym | Sodium dichloroisocyanurate dihydrate |
PubChem CID | 129893375 |
IUPAC Name | 1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium;dihydrate |
SMILES | C1(=O)NC(=O)N(C(=O)N1Cl)Cl.O.O.[Na] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11946581
|
Thermo Scientific Acros
436450500 |
50 g | Glass bottle |
34.77 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11300290
|
Thermo Scientific Acros
436452500 |
250 g | Glass bottle |
53.77 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11936581
|
Thermo Scientific Acros
436450010 |
1 kg | Glass bottle |
163.82 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
52671-45-1 | |
237.96 | |
Sodium dichloroisocyanurate dihydrate | |
1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium;dihydrate |
C3Cl2N3NaO3·H2O | |
FIHQYHSNTSJNGG-UHFFFAOYSA-N | |
129893375 | |
C1(=O)NC(=O)N(C(=O)N1Cl)Cl.O.O.[Na] |
Specifications
240.0°C to 250.0°C | |
White | |
96.0 to 104% (on dry substance) (Iodometry) | |
Sodium dichloroisocyanurate dihydrate | |
FIHQYHSNTSJNGG-UHFFFAOYSA-N | |
1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium;dihydrate | |
129893375 | |
97% | |
15% max. |
52671-45-1 | |
Crystals or Granules | |
C3Cl2N3NaO3·H2O | |
Solubility in water: ca 250g/L (25°C) | |
C1(=O)NC(=O)N(C(=O)N1Cl)Cl.O.O.[Na] | |
237.96 | |
237.96 | |
Authentic | |
Dichloroisocyanuric acid sodium salt monohydrate |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Contact with acids liberates t
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major
GHS Signal Word: Danger
- Dichloroisocyanuric acid sodium salt dihydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.