missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodifluoromethane, SPEX CertiPrep™

Product Code. 13545130
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Dichlorodifluoromethane75-71-81000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 75-71-8, 67-56-1
Packaging 2mL amber prescored glass ampule
CAS Min % 0.1, 99.9
Quantity 1 mL
CAS Max % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Molecular Formula CCl2F2
InChI Key PXBRQCKWGAHEHS-UHFFFAOYSA-N
SMILES C(F)(F)(Cl)Cl
IUPAC Name dichloro(difluoro)methane
Molecular Weight (g/mol) 120.908
PubChem CID 6391
Solvent or Matrix Methanol-P&T
Chemical Name or Material Dichlorodifluoromethane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dichlorodifluoromethane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
Supplemental information
  • MIXTURE LIST-Contains : methanol, dichlorodifluoromethane

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'