Learn More
Dibutyl phosphite, 14.5-16% P, Thermo Scientific Chemicals
30.30 € - 92.10 €
Chemical Identifiers
CAS | 1809-19-4 |
---|---|
Molecular Formula | C8H19O3P |
Molecular Weight (g/mol) | 194.21 |
MDL Number | MFCD00066633 |
InChI Key | OSPSWZSRKYCQPF-UHFFFAOYSA-N |
Synonym | dibutyl phosphite, dibutyl phosphonate, phosphonic acid, dibutyl ester, di-n-butylphosphite, dibutoxyphosphine oxide, mobil dbhp, dibutyl hydrogen phosphonate, di-n-butyl hydrogen phosphite, dibutylfosfit, phosphorous acid, dibutyl ester |
PubChem CID | 6327349 |
IUPAC Name | dibutoxy(oxo)phosphanium |
SMILES | CCCCO[P+](=O)OCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10573701
|
Thermo Scientific Acros
155061000 |
100 g | Glass bottle |
30.30 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10337520
|
Thermo Scientific Acros
155065000 |
500 g | Glass bottle |
92.10 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1809-19-4 | |
194.21 | |
OSPSWZSRKYCQPF-UHFFFAOYSA-N | |
6327349 | |
CCCCO[P+](=O)OCCCC |
C8H19O3P | |
MFCD00066633 | |
dibutyl phosphite, dibutyl phosphonate, phosphonic acid, dibutyl ester, di-n-butylphosphite, dibutoxyphosphine oxide, mobil dbhp, dibutyl hydrogen phosphonate, di-n-butyl hydrogen phosphite, dibutylfosfit, phosphorous acid, dibutyl ester | |
dibutoxy(oxo)phosphanium |
Specifications
1809-19-4 | |
118°C to 119°C (11.0 mmHg) | |
Authentic | |
C8H19O3P | |
1.4225 to 1.4245 | |
01, I, 187 | |
dibutyl phosphite, dibutyl phosphonate, phosphonic acid, dibutyl ester, di-n-butylphosphite, dibutoxyphosphine oxide, mobil dbhp, dibutyl hydrogen phosphonate, di-n-butyl hydrogen phosphite, dibutylfosfit, phosphorous acid, dibutyl ester | |
OSPSWZSRKYCQPF-UHFFFAOYSA-N | |
dibutoxy(oxo)phosphanium | |
6327349 | |
14.5 to 16% (P) | |
Dibutyl phosphite, 96% |
0.9950g/mL | |
121°C | |
96% | |
[CH3(CH2)3O]2P(O)H | |
MFCD00066633 | |
0.995 | |
Solubility in water: slightly soluble | |
CCCCO[P+](=O)OCCCC | |
194.21 | |
194.21 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 217-316-1
missing translation for 'rtecsNumber' : HS6475000
missing translation for 'tsca' : TSCA
- Dibutyl phosphite
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only