Learn More
Dibromomethane, 99%, Thermo Scientific Chemicals
41.51 € - 320.60 €
Chemical Identifiers
CAS | 74-95-3 |
---|---|
Molecular Formula | CH2Br2 |
Molecular Weight (g/mol) | 173.83 |
MDL Number | MFCD00000168 |
InChI Key | FJBFPHVGVWTDIP-UHFFFAOYSA-N |
Synonym | methylene bromide, methane, dibromo, methylene dibromide, dibromomethylene, rcra waste number u068, dibrommethan, methylenbromid, dibromo-methane, ccris 939, rcra waste no. u068 |
PubChem CID | 3024 |
ChEBI | CHEBI:47077 |
IUPAC Name | dibromomethane |
SMILES | C(Br)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10615532
|
Thermo Scientific Acros
112831000 |
100 mL | Glass bottle |
41.51 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10224570
|
Thermo Scientific Acros
112835000 |
500 mL | Glass bottle |
106.53 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10124523
|
Thermo Scientific Acros
112830010 |
1 L | Glass bottle |
161.81 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11984481
|
Thermo Scientific Acros
112830025 |
2.5 L | Glass Bottle |
320.60 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
74-95-3 | |
173.83 | |
FJBFPHVGVWTDIP-UHFFFAOYSA-N | |
3024 | |
dibromomethane |
CH2Br2 | |
MFCD00000168 | |
methylene bromide, methane, dibromo, methylene dibromide, dibromomethylene, rcra waste number u068, dibrommethan, methylenbromid, dibromo-methane, ccris 939, rcra waste no. u068 | |
CHEBI:47077 | |
C(Br)Br |
Specifications
74-95-3 | |
Colorless to Brown | |
96.0°C to 98.0°C | |
98.5% min. (GC) | |
CH2Br2 | |
MFCD00000168 | |
11,337; 12,322; 16,393 | |
2.47 | |
Solubility in water: 1g/L (20°C). Other solubilities: miscible with alcohol, ether, acetone and, chloroform | |
C(Br)Br | |
173.83 | |
CHEBI:47077 | |
99% | |
Dibromomethane, 99% |
-52.0°C | |
2.4700g/mL | |
Authentic | |
CH2Br2 | |
1.5400 to 1.5420 | |
01,67 | |
15,6133 | |
methylene bromide, methane, dibromo, methylene dibromide, dibromomethylene, rcra waste number u068, dibrommethan, methylenbromid, dibromo-methane, ccris 939, rcra waste no. u068 | |
FJBFPHVGVWTDIP-UHFFFAOYSA-N | |
dibromomethane | |
3024 | |
173.83 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful to aquatic life with long lasting effects.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-824-2
missing translation for 'rtecsNumber' : PA7350000
missing translation for 'tsca' : TSCA
- Dibromomethane
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P273-Avoid release to the environment.
- P501-Dispose of contents/container to an approved waste disposal plant.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.