missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dibenzoyl-L-tartaric acid,98%, Thermo Scientific Chemicals

Product Code. 11423257
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(-)-Dibenzoyl-L-tartaric acid, anhydrous is used as a reagent used to produce chiral salts. Also used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Dibenzoyl-L-tartaric acid, anhydrous is used as a reagent used to produce chiral salts. Also used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
It is sensitive to moisture. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2743-38-6
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00003074
InChI Key YONLFQNRGZXBBF-ZIAGYGMSSA-N
Synonym dibenzoyl-l-tartaric acid, 2r,3r-2,3-bis benzoyloxy succinic acid, dibenzoyltartaric acid, --dibenzoyl-l-tartaric acid, dibenzoyl-l-tartaricacid, l-dibenzoyl tartaric acid, butanedioic acid, 2,3-bis benzoyloxy-, 2r,3r, unii-3q14b666vm, 2,3-bis benzoyloxy tartaric acid, o,o-dibenzoyl-+-tartaric acid
PubChem CID 75969
SMILES OC(=O)[C@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 154°C to 156°C
Solubility Information Slightly soluble in water.
IUPAC Name (2R,3R)-2,3-dibenzoyloxybutanedioic acid
Formula Weight 358.31
Percent Purity 99%
Quantity 500 g
Sensitivity Hygroscopic
Chemical Name or Material (-)-Dibenzoyl-L-tartaric acid, anhydrous

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'