missing translation for 'onlineSavingsMsg'
Learn More

Di-Sodium Hydrogen Orthophosphate Dodecahydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Di-Sodium Hydrogen Orthophosphate Dodecahydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical, Quantity: 500g, CAS: 10039-32-4, Melting Point: 35 deg.C, Molecular Formula: H25Na2O16P, Molecular Weight: 358.137, Packaging: Plastic powder jar

76.85 € - 77.25 €

Chemical Identifiers

CAS 10039-32-4
Molecular Formula H25Na2O16P
Molecular Weight (g/mol) 358.137
MDL Number 149181
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
Synonym disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
PubChem CID 61456
IUPAC Name disodium;hydrogen phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10102880
View Documents Promotion Details
Fisher Chemical
S/4400/53
500 g
77.25 €
500g
Please to purchase this item. Need a web account? Register with us today!
10204020
View Documents Promotion Details
Fisher Chemical
S/4400/60
1 kg
76.85 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Di-Sodium Hydrogen Orthophosphate Dodecahydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical, Quantity: 500g, CAS: 10039-32-4, Melting Point: 35 deg.C, Molecular Formula: H25Na2O16P, Molecular Weight: 358.137, Packaging: Plastic powder jar

Specifications

Chemical Identifiers

10039-32-4
358.137
DGLRDKLJZLEJCY-UHFFFAOYSA-L
61456
O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
H25Na2O16P
149181
disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
disodium;hydrogen phosphate;dodecahydrate

Specifications

35°C
9.0 to 9.4
H25Na2O16P
disodium phosphate dodecahydrate, disodium hydrogen phosphate dodecahydrate, phosphoric acid, disodium salt, dodecahydrate, di-sodium hydrogen phosphate dodecahydrate, sodium hydrogenphosphate dodecahydrate, unii-e1w4n241fo, sodium phosphate dibasic dodecahydrate, sodium phosphate, dibasic, dodecahydrate, disodium orthophosphate, dodecahydrate, sodium monohydrogen phosphate dodecahydrate 2:1:1:12
O.O.O.O.O.O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
358.137
358.13g/mol
10039-32-4
Solid
149181
DGLRDKLJZLEJCY-UHFFFAOYSA-L
disodium;hydrogen phosphate;dodecahydrate
61456
Plastic powder jar
Videos
SDS
Documents

Documents

Product Certifications