missing translation for 'onlineSavingsMsg'
Learn More

Di-Sodium Hydrogen Orthophosphate Anhydrous, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10419190
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar

HNa2O4P, CAS Number-7558-79-4, disodium orthophosphate, acetest, disodium hydrogen phosphate, disodium hydrogenorthophosphate, disodium phosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt, soda phosphate, sodium phosphate dibasic, dibasic sodium phosphate, 3kg, CHEBI:34683, White

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Color White
pH 8.7 to 9.3
Physical Form Powder/Solid
Quantity 5 kg
IUPAC Name disodium;hydrogen phosphate
Formula Weight 141.96g/mol
Packaging Plastic keg
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'