Learn More
Di-n-hexylamine, 98+%, Thermo Scientific Chemicals
44.00 € - 121.00 €
Chemical Identifiers
CAS | 143-16-8 |
---|---|
Molecular Formula | C12H27N |
Molecular Weight (g/mol) | 185.355 |
MDL Number | MFCD00009521 |
InChI Key | PXSXRABJBXYMFT-UHFFFAOYSA-N |
Synonym | dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine |
PubChem CID | 8920 |
IUPAC Name | N-hexylhexan-1-amine |
SMILES | CCCCCCNCCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11312466
|
Thermo Scientific Alfa Aesar
L06326.AE |
100 mL |
44.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11322466
|
Thermo Scientific Alfa Aesar
L06326.AP |
500 mL |
121.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Di-n-hexylamine is used in organic synthesis and involved in the preparation of succinic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDi-n-hexylamine is used in organic synthesis and involved in the preparation of succinic acid.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides and acid anhydrides.
Chemical Identifiers
143-16-8 | |
185.355 | |
PXSXRABJBXYMFT-UHFFFAOYSA-N | |
8920 | |
CCCCCCNCCCCCC |
C12H27N | |
MFCD00009521 | |
dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine | |
N-hexylhexan-1-amine |
Specifications
143-16-8 | |
0.79 | |
104°C (219°F) | |
1.4335 | |
UN2927 | |
dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine | |
PXSXRABJBXYMFT-UHFFFAOYSA-N | |
N-hexylhexan-1-amine | |
8920 | |
≥98% |
3°C | |
236°C | |
C12H27N | |
MFCD00009521 | |
1738519 | |
Miscible with water. | |
CCCCCCNCCCCCC | |
185.355 | |
185.36 | |
Di-n-hexylamine |
Safety and Handling
GHS H Statement
H310-H330-H314-H318-H302
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P261-P262-P264b-P270-P271-P280-P285-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P363-P501c
H301-H310+H330-H314
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 205-588-4
missing translation for 'rtecsNumber' : IH6600000
missing translation for 'tsca' : Yes
RUO – Research Use Only