missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-hexyl disulfide, tech. 85%, Thermo Scientific Chemicals
39.80 € - 140.00 €
Chemical Identifiers
CAS | 10496-15-8 |
---|---|
Molecular Formula | C12H26S2 |
Molecular Weight (g/mol) | 234.46 |
MDL Number | MFCD00027300 |
InChI Key | GJPDBURPGLWRPW-UHFFFAOYSA-N |
Synonym | hexyl disulfide, dihexyl disulfide, di-n-hexyl disulfide, disulfide, dihexyl, dihexyl disulphide, 1-hexyldisulfanyl hexane, disulfide,dihexyl, dihexyl persulfide, n-hexyl disulfide, di-n-hexyldisulfide |
PubChem CID | 82675 |
IUPAC Name | 1-(hexyldisulfanyl)hexane |
SMILES | CCCCCCSSCCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11425147
|
Thermo Scientific Alfa Aesar
B22568.06 |
5 g |
39.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11435147
|
Thermo Scientific Alfa Aesar
B22568.14 |
25 g |
140.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10496-15-8 | |
234.46 | |
GJPDBURPGLWRPW-UHFFFAOYSA-N | |
82675 | |
CCCCCCSSCCCCCC |
C12H26S2 | |
MFCD00027300 | |
hexyl disulfide, dihexyl disulfide, di-n-hexyl disulfide, disulfide, dihexyl, dihexyl disulphide, 1-hexyldisulfanyl hexane, disulfide,dihexyl, dihexyl persulfide, n-hexyl disulfide, di-n-hexyldisulfide | |
1-(hexyldisulfanyl)hexane |
Specifications
10496-15-8 | |
>110°C (230°F) | |
C12H26S2 | |
hexyl disulfide, dihexyl disulfide, di-n-hexyl disulfide, disulfide, dihexyl, dihexyl disulphide, 1-hexyldisulfanyl hexane, disulfide,dihexyl, dihexyl persulfide, n-hexyl disulfide, di-n-hexyldisulfide | |
CCCCCCSSCCCCCC | |
234.46 | |
234.47 | |
Technical |
0.919 | |
Unpleasant | |
MFCD00027300 | |
GJPDBURPGLWRPW-UHFFFAOYSA-N | |
1-(hexyldisulfanyl)hexane | |
82675 | |
85% | |
Di-n-hexyl disulfide |
Safety and Handling
missing translation for 'einecsNumber' : 234-022-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only