Learn More
Di-n-butyltin oxide, 98%, Thermo Scientific Chemicals
17.00 € - 94.50 €
Chemical Identifiers
CAS | 818-08-6 |
---|---|
Molecular Formula | C8H18OSn |
Molecular Weight (g/mol) | 248.941 |
MDL Number | MFCD00001992 |
InChI Key | JGFBRKRYDCGYKD-UHFFFAOYSA-N |
Synonym | dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin |
PubChem CID | 61221 |
IUPAC Name | dibutyl(oxo)tin |
SMILES | CCCC[Sn](=O)CCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11363546
|
Thermo Scientific Alfa Aesar
L14491.14 |
25 g | |||||||
11373546
|
Thermo Scientific Alfa Aesar
L14491.22 |
100 g | |||||||
11383546
|
Thermo Scientific Alfa Aesar
L14491.36 |
500 g |
94.50 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
818-08-6 | |
248.941 | |
JGFBRKRYDCGYKD-UHFFFAOYSA-N | |
61221 | |
CCCC[Sn](=O)CCCC |
C8H18OSn | |
MFCD00001992 | |
dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin | |
dibutyl(oxo)tin |
Specifications
818-08-6 | |
Odorless | |
[CH3(CH2)3]2SnO | |
UN3146 | |
dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin | |
CCCC[Sn](=O)CCCC | |
248.941 | |
248.94 | |
Di-n-butyltin oxide |
>300°C | |
C8H18OSn | |
MFCD00001992 | |
4126243 | |
JGFBRKRYDCGYKD-UHFFFAOYSA-N | |
dibutyl(oxo)tin | |
61221 | |
98% |
Safety and Handling
GHS H Statement
H300-H319
Fatal if swallowed.
Causes serious eye irritation.
GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
missing translation for 'rtecsNumber' : WH7175000
missing translation for 'tsca' : TSCA
RUO – Research Use Only