missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyltin bis(2-ethylhexanoate), Thermo Scientific Chemicals

Product Code. 11324819
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Di-n-butyltin bis(2-ethylhexanoate) is used in silanol condensation reactions for caulk and sealant applications and in the production of silicones. It is also used in one or two component systems which are compatible with aromatic solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2781-10-4
MDL Number MFCD00015261

Specifications

Melting Point 57°C to 59°C
Density 1.070 g/mL
Boiling Point 220°C (2 mmHg)
Odor Odorless
Molecular Formula C24H48O4Sn
Quantity 25 g
UN Number UN3146
Synonym dibutyltin bis 2-ethylhexanoate, dibutyltin di 2-ethylhexoate, stannane, dibutylbis 2-ethyl-1-oxohexyl oxy, di-n-butyltin di-2-ethylhexanoate, di-n-butyltin bis 2-ethylhexanoate, dibutyltin di-2-ethylhexanoate, dibutyltin di 2-ethylhexanoate, dibutylbis 2-ethyl-1-oxohexyl oxy stannane, tin, dibutyl-, di 2-ethylhexoate, dibutyltin di-2-ethylhexoate
Solubility Information Soluble in mineral oil, acetone, xylene
InChI Key GPDWNEFHGANACG-UHFFFAOYNA-L
SMILES CCCCC(CC)C(=O)O[Sn](CCCC)(CCCC)OC(=O)C(CC)CCCC
IUPAC Name dibutyl[(2-ethylhexanoyl)oxy]stannyl 2-ethylhexanoate
Molecular Weight (g/mol) 519.35
Formula Weight 519.34
Physical Form Lumps
Chemical Name or Material Di-n-butyltin bis (2-ethylhexanoate)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.