missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyltin bis(2,4-pentanedionate), 95%, Thermo Scientific Chemicals

Product Code. 11416786
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22673-19-4
Molecular Formula C18H32O4Sn
MDL Number MFCD00077994
Synonym dibutyltin bis acetylacetonate, di-n-butylbis 2,4-pentanedionate tin, dibutyltin bis 2,4-pentanedionate, 3-penten-2-one, 4,4'-dibutylstannylene bis oxy bis-, 3z,3'z, 3z-4-dibutyl 2z-4-oxopent-2-en-2-yl oxy stannyl oxy pent-3-en-2-one, z-4-dibutyl-z-1-methyl-3-oxo-but-1-enoxy stannyl oxypent-3-en-2-one

Specifications

Melting Point 26°C
Density 1.22
Boiling Point 150°C
Flash Point 40°C (104°F)
Odor Weak
Refractive Index 1.515
Quantity 1000 g
UN Number UN1992
Formula Weight 431.16
Percent Purity 95%
Chemical Name or Material Di-n-butyltin bis(2,4-pentanedionate)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'