missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-butyl sulfoxide, 97%, Thermo Scientific Chemicals
34.87 € - 113.57 €
Chemical Identifiers
CAS | 2168-93-6 |
---|---|
Molecular Formula | C8H18OS |
Molecular Weight (g/mol) | 162.291 |
MDL Number | MFCD00002093 |
InChI Key | LOWMYOWHQMKBTM-UHFFFAOYSA-N |
Synonym | dibutyl sulfoxide, butyl sulfoxide, n-butyl sulfoxide, dibutyl sulphoxide, di-n-butyl sulfoxide, butane, 1,1'-sulfinylbis, sulfoxide, dibutyl, di-n-butylsulfoxide, butane,1-butylsulfinyl, butylsulfinyl butane |
PubChem CID | 16568 |
IUPAC Name | 1-butylsulfinylbutane |
SMILES | CCCCS(=O)CCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460078
|
Thermo Scientific Alfa Aesar
B23672.06 |
5 g |
34.87 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470078
|
Thermo Scientific Alfa Aesar
B23672.14 |
25 g |
113.57 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as an active pharmaceutical intermediate and in the extraction of thorium(IV) from nitric acid solutions in xylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an active pharmaceutical intermediate and in the extraction of thorium(IV) from nitric acid solutions in xylene.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
Chemical Identifiers
2168-93-6 | |
162.291 | |
LOWMYOWHQMKBTM-UHFFFAOYSA-N | |
16568 | |
CCCCS(=O)CCCC |
C8H18OS | |
MFCD00002093 | |
dibutyl sulfoxide, butyl sulfoxide, n-butyl sulfoxide, dibutyl sulphoxide, di-n-butyl sulfoxide, butane, 1,1'-sulfinylbis, sulfoxide, dibutyl, di-n-butylsulfoxide, butane,1-butylsulfinyl, butylsulfinyl butane | |
1-butylsulfinylbutane |
Specifications
2168-93-6 | |
0.832 | |
121°C (249°F) | |
1.467 | |
1747425 | |
dibutyl sulfoxide, butyl sulfoxide, n-butyl sulfoxide, dibutyl sulphoxide, di-n-butyl sulfoxide, butane, 1,1'-sulfinylbis, sulfoxide, dibutyl, di-n-butylsulfoxide, butane,1-butylsulfinyl, butylsulfinyl butane | |
LOWMYOWHQMKBTM-UHFFFAOYSA-N | |
1-butylsulfinylbutane | |
16568 | |
97% |
31°C to 34°C | |
250°C | |
C8H18OS | |
MFCD00002093 | |
Hygroscopic | |
Slightly soluble in water. | |
CCCCS(=O)CCCC | |
162.291 | |
162.3 | |
Di-n-butyl sulfoxide |
Safety and Handling
missing translation for 'einecsNumber' : 218-511-4
missing translation for 'rtecsNumber' : EK5700000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only