missing translation for 'onlineSavingsMsg'
Learn More

Di(2-pyridyl) ketone, 98%, Thermo Scientific Chemicals

Product Code. 15411548
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

di-2-Pyridyl ketone has been used in the preparation of unusual {Ni(II)(3)Ln(III)(?-OR)(6)}(3+) complexes with a star topology. Its monoanionic form has been used in the synthesis of triangular Ni(2)M (M = lanthanide, Y) complexes. The Schiff base ligands derived from di-2-pyridyl ketone exhibit ant proliferative activity in SK-N-MC neuroepithelioma (cancer) cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
di-2-Pyridyl ketone has been used in the preparation of unusual {Ni(II)(3)Ln(III)(μ-OR)(6)}(3+) complexes with a ″star″ topology. Its monoanionic form has been used in the synthesis of triangular Ni(2)M (M = lanthanide, Y) complexes. The Schiff base ligands derived from di-2-pyridyl ketone exhibit ant proliferative activity in SK-N-MC neuroepithelioma (cancer) cells.

Solubility
Low solubility in water.

Notes
Desiccate at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19437-26-4
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.198
MDL Number MFCD00006288
InChI Key QPOWUYJWCJRLEE-UHFFFAOYSA-N
Synonym di-2-pyridyl ketone, di pyridin-2-yl methanone, bis 2-pyridyl ketone, 2,2'-carbonyldipyridine, di 2-pyridyl ketone, methanone, di-2-pyridinyl, 2,2'-dipyridyl ketone, 2-pyridine-2-carbonyl pyridine, pyridyl ketone
PubChem CID 88065
IUPAC Name dipyridin-2-ylmethanone
SMILES C1=CC=NC(=C1)C(=O)C2=CC=CC=N2

Specifications

Melting Point 52°C to 55°C
Flash Point 110°C (230°F)
Quantity 5 g
Beilstein 126598
Solubility Information Low solubility in water.
Formula Weight 184.2
Percent Purity 99%
Chemical Name or Material Di(2-pyridyl) ketone
compliance-icons
Product Identifier
  • Di(2-pyridyl) ketone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Bis(2-pyridyl) ketone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'