Learn More
Desvenlafaxine, 97%, Thermo Scientific Chemicals
51.56 € - 132.66 €
Chemical Identifiers
CAS | 93413-62-8 |
---|---|
Molecular Formula | C16H25NO2 |
Molecular Weight (g/mol) | 263.38 |
InChI Key | KYYIDSXMWOZKMP-UHFFFAOYSA-N |
Synonym | desvenlafaxine, o-desmethylvenlafaxine, 4-2-dimethylamino-1-1-hydroxycyclohexyl ethyl phenol, pristiq, o-desmethyl venlafaxine, desvenlafaxine inn:ban, desvenlafaxine inn, o-desmethylvenlafaxine odv, 1-2-dimethylamino-1-4-hydroxyphenyl ethyl cyclohexanol |
PubChem CID | 125017 |
ChEBI | CHEBI:83527 |
IUPAC Name | 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol |
SMILES | CN(C)CC(C1=CC=C(C=C1)O)C2(CCCCC2)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15668745
|
Thermo Scientific Acros
461570010 |
1 g |
51.56 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15678745
|
Thermo Scientific Acros
461570050 |
5 g |
132.66 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
93413-62-8 | |
263.38 | |
desvenlafaxine, o-desmethylvenlafaxine, 4-2-dimethylamino-1-1-hydroxycyclohexyl ethyl phenol, pristiq, o-desmethyl venlafaxine, desvenlafaxine inn:ban, desvenlafaxine inn, o-desmethylvenlafaxine odv, 1-2-dimethylamino-1-4-hydroxyphenyl ethyl cyclohexanol | |
CHEBI:83527 | |
CN(C)CC(C1=CC=C(C=C1)O)C2(CCCCC2)O |
C16H25NO2 | |
KYYIDSXMWOZKMP-UHFFFAOYSA-N | |
125017 | |
4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol |
Specifications
93413-62-8 | |
Authentic | |
Glass Bottle | |
desvenlafaxine, o-desmethylvenlafaxine, 4-2-dimethylamino-1-1-hydroxycyclohexyl ethyl phenol, pristiq, o-desmethyl venlafaxine, desvenlafaxine inn:ban, desvenlafaxine inn, o-desmethylvenlafaxine odv, 1-2-dimethylamino-1-4-hydroxyphenyl ethyl cyclohexanol | |
CN(C)CC(C1=CC=C(C=C1)O)C2(CCCCC2)O | |
263.38 | |
CHEBI:83527 | |
97% |
0.5% max. | |
97.5% min. (HPLC) | |
C16H25NO2 | |
KYYIDSXMWOZKMP-UHFFFAOYSA-N | |
4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol | |
125017 | |
263.38 | |
Desvenlafaxine |
Safety and Handling
GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. If eye irritation persists: Get medical advice/attention. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
- Desvenlafaxine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Desvenlafaxine
RUO – Research Use Only