missing translation for 'onlineSavingsMsg'
Learn More

Deoxyribonucleic Acid Sodium Salt, Fisher BioReagents

51.60 € - 787.00 €

Chemical Identifiers

CAS 68938-01-2
Molecular Formula C17H15BrFNO3
Molecular Weight (g/mol) 50000-100000
MDL Number MFCD00130922
InChI Key GXZCYECDYOBPGH-UHFFFAOYNA-N
PubChem CID 73904662
IUPAC Name (2R)-3-(3-bromophenyl)-2-[[2-(4-fluorophenyl)acetyl]amino]propanoic acid
SMILES *
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10502955
View Documents
Fisher BioReagents
BP2514-250
250 mg Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11403074
View Documents
Fisher BioReagents
BP2514-1
1 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10428923
View Documents
Fisher BioReagents
BP2514-5
5 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11361126
View Documents
Fisher BioReagents
BP2514-10
10 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Deoxyribonucleic Acid Sodium Salt is used in several biochemical research applications.
Specifications

Chemical Identifiers

68938-01-2
50000-100000
GXZCYECDYOBPGH-UHFFFAOYNA-N
(2R)-3-(3-bromophenyl)-2-[[2-(4-fluorophenyl)acetyl]amino]propanoic acid
C17H15BrFNO3
MFCD00130922
73904662
*

Specifications

68938-01-2
14% max.
MFCD00130922
*
50000-100000
6000000
Deoxyribonucleic Acid Sodium Salt, From Salmon Sperm
White
C17H15BrFNO3
GXZCYECDYOBPGH-UHFFFAOYNA-N
(2R)-3-(3-bromophenyl)-2-[[2-(4-fluorophenyl)acetyl]amino]propanoic acid
73904662
Solid
Videos
Safety and Handling

Safety and Handling

Recommended Storage : 4°C

Product Identifier
  • Deoxyribonucleic Acid, Sodium Salt

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.