missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Deoxyribonucleic Acid Sodium Salt
69.55 € - 198.99 €
Chemical Identifiers
CAS | 68938-01-2 |
---|---|
Molecular Formula | C17H15BrFNO3 |
Molecular Weight (g/mol) | 50000-100000 |
MDL Number | MFCD00130922 |
InChI Key | GXZCYECDYOBPGH-UHFFFAOYNA-N |
Synonym | DNA sodium salt |
PubChem CID | 73904662 |
IUPAC Name | (2R)-3-(3-bromophenyl)-2-[[2-(4-fluorophenyl)acetyl]amino]propanoic acid |
SMILES | * |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10762012
|
Thermo Scientific Chemicals
406250250 |
25 g | Glass bottle |
69.55 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10598122
|
Thermo Scientific Chemicals
406251000 |
100 g | Glass bottle |
198.99 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68938-01-2 | |
50000-100000 | |
GXZCYECDYOBPGH-UHFFFAOYNA-N | |
73904662 | |
* |
C17H15BrFNO3 | |
MFCD00130922 | |
DNA sodium salt | |
(2R)-3-(3-bromophenyl)-2-[[2-(4-fluorophenyl)acetyl]amino]propanoic acid |
Specifications
68938-01-2 | |
Powder | |
MFCD00130922 | |
GXZCYECDYOBPGH-UHFFFAOYNA-N | |
(2R)-3-(3-bromophenyl)-2-[[2-(4-fluorophenyl)acetyl]amino]propanoic acid | |
73904662 | |
Authentic | |
Deoxyribonucleic acid sodium salt |
White to Tan | |
C17H15BrFNO3 | |
DNA sodium salt | |
* | |
50000-100000 | |
Pure | |
18% max. |
Safety and Handling
Product Identifier
- Deoxyribonucleic acid sodium salt, M.W. 50,000-100,000 Dalton
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title