missing translation for 'onlineSavingsMsg'
Learn More

Deoxycholic Acid Sodium Salt, Fisher BioReagents

Product Code. 10346653
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

For solubilizing and isolating membrane-bound proteins in an active state

This anionic detergent is useful for solubilizing and isolating membrane-bound proteins in an active state.
TRUSTED_SUSTAINABILITY

Specifications

Color White
CAS 302-95-4
Physical Form Powder/Solid
Melting Point 357°C
pH 7.5 to 10
Quantity 100 g
Identification Pass Test
Packaging Poly Bottle
Assay Percent Range ≥99 %
Molecular Formula C24H39NaO4
MDL Number MFCD00064139
Synonym Sodium Deoxycholate, 3-alpha, 12-alpha-Dihydroxy-5-beta-cholan-24-oic acid, sodium salt
InChI Key FHHPUSMSKHSNKW-SMOYURAASA-M
SMILES [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])CC[C@]2([H])C[C@H](O)CC[C@]12C
IUPAC Name sodium;(4R)-4-[(3R,5R,8R,10S,12S,13R,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
Molecular Weight (g/mol) 414.56
PubChem CID 91810855
Formula Weight 414.58
Percent Purity ≥99%
Chemical Name or Material Deoxycholic Acid Sodium Salt
Show More Show Less
compliance-icons
Product Identifier
  • Deoxycholic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'