missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Density Standards (15 °C), Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C
Brand: Paragon Scientific DEN15-09
This item is not returnable.
View return policy
Description
- Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in accordance with primary level ASTM D1480 methodology
- Ensures full compliance to ASTM and IP test method protocols.
- Supplied in tamper evident packaging with a 2 year shelf life.
- Fully traceable to international standards.
- Supplied with MSDS.
- Tamper evident security packaging.
- Manufactured in the United Kingdom
Specifications
60 mL | |
0.9823 g/mL (15°C) | |
Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
Nominal values quoted