Learn More
Density Standard (20°C); UKAS ISO/IEC17025 and ISO 17034 certified, nominal density 0.8708 g/mL (20°C), 500 mL, Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C to 150°C
Brand:  Paragon Scientific DEN20-06-500ML
Product Code. 16060120
  • 484.00 € / 500mL
Explore more special offers
Description
Description
  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols
  • Supplied in tamper evident packaging with a 2 year shelf life
  • Manufactured in the United Kingdom
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Nominal values quoted. Certified Values will vary from batch to batch.